Merck
  • Polyamidoamine dendrimer grafted forward osmosis membrane with superior ammonia selectivity and robust antifouling capacity for domestic wastewater concentration.
Polyamidoamine dendrimer grafted forward osmosis membrane with superior ammonia selectivity and robust antifouling capacity for domestic wastewater concentration.
Water research (2019-01-27)
Xian Bao, Qinglian Wu, Wenxin Shi, Wei Wang, Huarong Yu, Zhigao Zhu, Xinyu Zhang, Zhiqiang Zhang, Ruijun Zhang, Fuyi Cui
ZUSAMMENFASSUNG
Developing a forward osmosis (FO) membrane with superior ammonia selectivity and robust antifouling performance is important for treating domestic wastewater (DW) but challenging due to the similar polarities and hydraulic radii of NH4+ and water molecules. Herein, we investigated the feasibility of using polyamidoamine (PAMAM) dendrimer to simultaneously enhance the ammonia rejection rate and antifouling capacity of the thin-film composite (TFC) FO membrane. PAMAM dendrimer with abundant, easily-protonated, terminal amine groups was grafted on TFC-FO membrane surface via covalent bonds, which inspired the TFC-FO membrane surface with appreciable Zeta potential (isoelectric point: pH = 5.5) and outstanding hydrophilicity (water contact angle: 39.83 ± 0.57°). Benefiting from the electrostatic repulsion between the protonated amine layer and NH4+-N as well as the concentration-induced diffusion resistance, the introduction of PAMAM dendrimer endowed the grafted membrane with a superior NH4+-N rejection rate of 98.23% and a significantly reduced the reverse solute flux when using NH4Cl solutions as feed solution. Meanwhile, the perfect balance between the electrostatic repulsion to positively-charged micromoleculer ions (metal ions and NH4+-N) and the electrostatic attraction to negatively-charged macromolecular organic foulants together with the hydrophilic nature of amine groups facilitated the enhancement of the grafted membranes in antifouling capacity and hence the NH4+-N selectivity (rejection rate of 91.81%) during the concentration of raw DW. The overall approach of this work opens up a frontier for preparation of ammonia-selective and antifouling TFC-FO membrane.