Merck
Ultrastructural Alteration of Pulmonary Capillary Endothelial Glycocalyx During Endotoxemia.
Chest (2018-03-21)
Risa Inagawa, Hideshi Okada, Genzou Takemura, Kodai Suzuki, Chihiro Takada, Hirohisa Yano, Yoshiaki Ando, Takahiro Usui, Yasuaki Hotta, Nagisa Miyazaki, Akiko Tsujimoto, Ryogen Zaikokuji, Atsumu Matsumoto, Tomonori Kawaguchi, Tomoaki Doi, Takahiro Yoshida, Shozo Yoshida, Keisuke Kumada, Hiroaki Ushikoshi, Izumi Toyoda, Shinji Ogura
ZUSAMMENFASSUNG
The most recent diagnostic criteria for sepsis include organ failure. Microvascular endothelial injury is believed to lead to the multiple organ failure seen in sepsis, although the precise mechanism is still controversial. ARDS is the primary complication during the sequential development of multiple organ dysfunction in sepsis, and endothelial injury is deeply involved. Sugar-protein glycocalyx coats all healthy vascular endothelium, and its disruption is one factor believed to contribute to microvascular endothelial dysfunction during sepsis. The goal of this study was to observe the three-dimensional ultrastructural alterations in the pulmonary capillary endothelium, including the glycocalyx, during sepsis-induced pulmonary vasculitis. This study investigated the three-dimensional ultrastructure of pulmonary vascular endothelial glycocalyx in a mouse lipopolysaccharide-induced endotoxemia model. Lungs were fixed with lanthanum-containing alkaline fixative to preserve the glycocalyx. On both scanning and transmission electron microscopic imaging, the capillary endothelial glycocalyx appeared as a moss-like structure entirely covering the endothelial cell surface in normal mice. In the septic lung following liposaccharide injection, however, this structure was severely disrupted; it appeared to be peeling away and coagulated. In addition, syndecan-1 levels were significantly reduced in the septic lung, and numerous spherical structures containing glycocalyx were observed on the endothelial surface. It appears that endothelial glycocalyx in the lung is markedly disrupted under experimental endotoxemia conditions. This finding supports the notion that disruption of the glycocalyx is causally related to the microvascular endothelial dysfunction that is characteristic of sepsis-induced ARDS.