Merck
  • Dual targeting of bromodomain-containing 4 by AZD5153 and BCL2 by AZD4320 against B-cell lymphomas concomitantly overexpressing c-MYC and BCL2.
Dual targeting of bromodomain-containing 4 by AZD5153 and BCL2 by AZD4320 against B-cell lymphomas concomitantly overexpressing c-MYC and BCL2.
Investigational new drugs (2018-06-23)
Tomoko Takimoto-Shimomura, Taku Tsukamoto, Saori Maegawa, Yuto Fujibayashi, Yayoi Matsumura-Kimoto, Yoshimi Mizuno, Yoshiaki Chinen, Yuji Shimura, Shinsuke Mizutani, Shigeo Horiike, Masafumi Taniwaki, Tsutomu Kobayashi, Junya Kuroda
ZUSAMMENFASSUNG
Despite the recent therapeutic progress, the prognoses of diffuse large B-cell lymphomas (DLBCLs) that concomitantly overexpress c-MYC and BCL2, i.e., double hit lymphoma (DHL) and double expressing lymphoma (DEL), remain poor. This study examined triple targeting of c-MYC, BCL2 and the B-cell receptor (BCR) signaling pathway for DHL and DEL. We first used AZD5153, a novel bivalent inhibitor for bromodomain-containing 4 (BRD4), in DHL- and DEL-derived cell lines, because BRD4 regulates disease type-oriented key molecules for oncogenesis. AZD5153 was more effective than conventional monovalent BRD4 inhibitors, JQ1 and I-BET151, in inhibiting cell proliferation of a DHL-derived cell line and two DEL-derived cell lines, with at least 10-fold lower half growth inhibitory concentrations. AZD5153 caused G1/S cell cycle blockade, while the apoptosis-inducing effect was relatively modest. At the molecular level, AZD5153 was potent in downregulating various molecules for oncogenesis, such as c-MYC, AKT2 and MAP3K; those involved in the BCR signaling pathway, such as CD19, BLNK and CD79B; and those associated with B-cell development, such as IKZF1, IKZF3, PAX5, POU2AF1 and EBF1. In contrast, AZD5153 did not decrease anti-apoptotic BCL2 proteins, and did not activate pro-apoptotic BH3-only proteins, except BAD. To augment cell death induction, we added a novel BH3-mimicking BCL2 inhibitor AZD4320 to AZD5153, and found that these two agents had a mostly synergistic antitumor effect by increasing cells undergoing apoptosis in all three cell lines. These results provide a rationale for dual targeting of BRD4 and BCL2 using AZD5153 and AZD4320 as a therapeutic strategy against DHL and DEL.