Merck
  • 8-OH-DPAT Induces Compulsive-like Deficit in Spontaneous Alternation Behavior: Reversal by MDMA but Not Citalopram.
8-OH-DPAT Induces Compulsive-like Deficit in Spontaneous Alternation Behavior: Reversal by MDMA but Not Citalopram.
ACS chemical neuroscience (2019-06-28)
Anna U Odland, Lea Jessen, Ciarán M Fitzpatrick, Jesper T Andreasen
ZUSAMMENFASSUNG
Rodents exhibit natural exploratory behaviors, which can be measured by the spontaneous alternation behavior (SAB) test. Perseverance in this test induced by the 5-hydroxytryptamine 1A receptor (5-HT1AR) agonist, 8-hydroxy-2-dipropylaminotetralin (8-OH-DPAT), resembles compulsive behaviors observed in humans and manifests as reduced alternation ratio. This study characterized 8-OH-DPAT-induced perseverance in the SAB test in C57BL/6JOlaHsd male mice by coadministration of WAY100635, citalopram and the 5-HT releasing agent, 3,4-methylenedioxymethamphetamine (MDMA), to deepen the understanding of 5-HT-dependent mechanisms. The 5-HT1AR mechanism of 8-OH-DPAT (1.0 mg/kg, p < 0.01) on perseverance was confirmed by coadministration of the 5-HT1AR antagonist, WAY100635 (2.0 mg/kg, p < 0.05), which attenuated the effects of 8-OH-DPAT. Such effects could also be reversed by MDMA (1.0 mg/kg, p < 0.05; 10.0 mg/kg, p < 0.001) but not citalopram. These findings confirm the importance of 5-HT in regulating perseverative behavior. Future investigations are required to determine the predictive validity of the 8-OH-DPAT-disrupted SAB test as an inducible mouse model of compulsivity.