Merck
Synergistic inhibition of pancreatic cancer with anti-PD-L1 and c-Myc inhibitor JQ1.
Oncoimmunology (2019-05-10)
Yu Pan, Qinglin Fei, Ping Xiong, Jianyang Yang, Zheyang Zhang, Xianchao Lin, Minggui Pan, Fengchun Lu, Heguang Huang
ZUSAMMENFASSUNG
Human pancreatic ductal adenocarcinoma (PDAC) exhibits marginal responses to anti-PD-1/PD-L1 immunotherapy and its mechanism remains poorly understood. We have investigated the effect of anti-PD-L1 and c-Myc inhibition in PDAC. Using 87 patients with PDAC from our hospital database we found a significant correlation between the expression of PD-L1 and c-Myc. Moreover, the expression of both PD-L1 and c-Myc was associated with poor overall survival. In addition, we confirmed this finding with the PDAC patients in the TCGA database. Using several PDAC cell lines we demonstrated a significant correlation between the expression of PD-L1 and c-Myc. We also found that expression of PD-L1 correlated with high-grade histology. JQ1, an inhibitor of c-Myc inhibited PD-L1 expression and tumor growth. Using xenograft models, we demonstrated that the combination of JQ1 and anti-PD-L1 antibody exerted synergistic inhibition of PDAC growth. Our data demonstrated that the expression of PD-L1 and c-Myc may be helpful prognostic biomarkers, and their inhibition may potentially serve as an effective treatment for PDAC.