Merck
  • Development and Evaluation of Optimized Thiolated Chitosan Proniosomal Gel Containing Duloxetine for Intranasal Delivery.
Development and Evaluation of Optimized Thiolated Chitosan Proniosomal Gel Containing Duloxetine for Intranasal Delivery.
AAPS PharmSciTech (2019-08-15)
Maryam Khatoon, Muhammad Farhan Sohail, Gul Shahnaz, Fiza Ur Rehman, Fakhar-Ud-Din, Asim Ur Rehman, Naseem Ullah, Umair Amin, Gul Majid Khan, Kifayat Ullah Shah
ZUSAMMENFASSUNG
Proniosomes offer excellent potential for improved drug delivery, through versatile routes, by overcoming the permeation barriers faced by several drugs. The study was aimed to develop a thiomer gel containing duloxetine proniosomes for the intranasal delivery, improving its bioavailability and brain delivery through olfactory system. Duloxetine-loaded proniosomes were optimized through Design-Expert Software, prepared by coacervation phase separation method and then characterized in vitro for different vesicle features, and permeation enhancement potential using various techniques. The formulation F2, out of all the trials, fulfilled the maximum requisite of highest entrapment efficiency (76.21 ± 1.24%) and minimum vesicle size (223.91 ± 11.07 nm). The F2 was embedded in thiolated chitosan gel rendering it mucoadhesive and further characterized. The in vitro release showed a sustained drug release from the mucoadhesive proniosomal gel with only 54% drug release as compared to that of 71% from proniosome over 8 h, following Higuchi drug release model. Ex vivo permeation studies showed the enhancement ratio for the mucoadhesive proniosomal gel to be 1.86-fold greater than proniosomes, indicating a significant improvement in transmucosal permeation. The results suggest that incorporation of proniosomes into thiolated gel can significantly improve its mucoadhesion and retention time in the nasal cavity for providing a sustained drug release. Thus, gel formulation could be considered as a promising approach for efficient intranasal drug delivery of duloxetine. Graphical Abstract.