Merck
  • Andrographolide binds to ATP-binding pocket of VEGFR2 to impede VEGFA-mediated tumor-angiogenesis.
Andrographolide binds to ATP-binding pocket of VEGFR2 to impede VEGFA-mediated tumor-angiogenesis.
Scientific reports (2019-03-13)
Kirti Kajal, Abir K Panda, Jyotsna Bhat, Dwaipayan Chakraborty, Sayantan Bose, Pushpak Bhattacharjee, Tania Sarkar, Subhrangsu Chatterjee, Santosh K Kar, Gaurisankar Sa
ZUSAMMENFASSUNG
Vasculogenesis and angiogenesis are process of formation of blood vessels. Blood vessels are evolved to distribute nutrients and oxygen to distant organs. These vessels are crucial for growth and repair of wounded tissue. During tumor condition there occurs imbalance in the growth of blood vessels which leads to neo-angiogenesis. Neo-angiogenesis is major perpetrator behind the establishment of tumor. Tumor cells secrete pro-angiogenic factor VEGFA which binds to VEGFR2 present over surface of endothelial cells and triggers formation of new blood vessels. To inhibit tumor-angiogenesis, a physiologically-safe small molecule inhibitor was screened which can potentially interact with kinase domain of VEGFR2 and inhibit its activity. Molecular-docking module and biochemical analysis identified andrographolide as one of the best docking molecules that binds to ATP-binding pocket of VEGFR2 and inhibits its kinase activity. Thus, for a more radical approach towards safe VEGFR2 inhibitor, andrographolide was repurposed to inhibit tumor-angiogenesis and reduce tumor burden.