Merck
  • Impact of angiotension I converting enzyme gene I/D polymorphism on running performance, lipid, and biochemical parameters in ultra-marathoners.
Impact of angiotension I converting enzyme gene I/D polymorphism on running performance, lipid, and biochemical parameters in ultra-marathoners.
Medicine (2019-07-25)
Yu-Hui Chiu, Jiun-I Lai, Chia-Ying Tseng, Shih-Hao Wang, Li-Hua Li, Wei-Fong Kao, Chorng-Kuang How, Wen-Han Chang, Chin-Yi Hsieh
ZUSAMMENFASSUNG
The insertion (I) or deletion (D) polymorphism in the angiotension I converting enzyme gene, (ACE I/D, rs1799752) is associated with human exercise endurance and performance. However, most of the aforementioned studies focus on marathons, swimming, and triathlons, while the ACE polymorphism in ultra-marathoners has not yet been reported. We studied the impact of ACE I/D polymorphism in ultra-marathoners and investigated its relationship with lipid profiles, interleukin-6 (IL-6), and high-sensitivity C-reactive protein (hs-CRP) levels in runners before and after ultra-marathon racing.This observational study used data from a 100-km ultra-marathon in Taipei, Taiwan. Twenty-four male participants were analyzed for their ACE insertion/deletion polymorphism, lipid profiles, hs-CRP, IL-6 in serum immediately before and after ultra-marathon running.In our 24 subjects analyzed, 7, 14, and 3 subjects were of I/I, I/D, and D/D genotypes, respectively. Runners with the D polymorphism (I/D and D/D) showed a trend of better performance in the 100-km ultra-marathon (measured by completion time in minutes, P = .036). In this group, the previous best marathon performance was also significantly better than the I/I group (P = .047). After adjusting for body mass index (BMI), the difference in performance was not significant. Ketone levels, IL-6, and hs-CRP levels were highly increased at immediately and 24-hour post-race. No correlation was found between different ACE polymorphisms and common biochemical parameters examined.We report the first study in the impact of the ACE I/D (rs1799752) on ultra-marathoners. Presence of the D polymorphism in ACE gene is associated with better performance, although the BMI of the runners contribute as a major factor. There was no difference in the biochemical or lipid parameters measured among different ACE polymorphisms.