Merck
  • Environmental media exert a bottleneck in driving the dynamics of antibiotic resistance genes in modern aquatic environment.
Environmental media exert a bottleneck in driving the dynamics of antibiotic resistance genes in modern aquatic environment.
Water research (2019-07-02)
Yihan Chen, Peng Li, Yuansheng Huang, Kaifeng Yu, Hongjie Chen, Kangping Cui, Qianli Huang, Junya Zhang, Karina Yew-Hoong Gin, Yiliang He
ZUSAMMENFASSUNG
With the rapid construction of dams worldwide, reservoir system has become a representation of modern aquatic environment. However, the profiles of antibiotic resistance genes (ARGs) and associated factor influencing their dynamics in modern aquatic environment (e.g., water phase, sediment phase, and soil phase) are largely unknown. Here, we comprehensively characterized the diversity, abundance, distribution of ARGs in a large drinking water reservoir using high-throughput quantitative PCR, as well as ranked the factors (e.g., mobile genetic elements (MGEs), bacteria community, bacterial biomass, antibiotics, and basic properties) influencing the profiles of ARGs on the basis of structural equation models (SEMs). Water phase was prone to harbor more diverse ARGs as compared to sediment phase and soil phase, and soil phase in drawdown area was a potential reservoir and hotspot for ARGs. Environmental media partially affected the ARG diversity in modern aquatic environment, while it observably influenced the distributions of ARGs and MGEs and their co-occurrence patterns. The pathways for the proliferation and spread of ARGs in water phase were both the horizontal gene transfer (HGT) and vertical gene transfer (VGT), while the dominant pathways in sediment phase and soil phase were the HGT and VGT, respectively. The SEMs demonstrated that MGEs contributed the most to drive the ARG dynamics in both water phase and sediment phase, while the most dominant factor for this in soil phase was bacterial community. Overall, environmental media exerted a bottleneck in driving the dynamics of ARGs in modern aquatic environment probably via diversifying the MGEs, bacterial community, bacterial biomass, antibiotics and basic properties.