Merck
Marine-derived n-3 fatty acids therapy for stroke.
The Cochrane database of systematic reviews (2019-06-27)
Celia Gabriela Alvarez Campano, Mary Joan Macleod, Lorna Aucott, Frank Thies
ZUSAMMENFASSUNG
Currently, with stroke burden increasing, there is a need to explore therapeutic options that ameliorate the acute insult. There is substantial evidence of a neuroprotective effect of marine-derived n-3 polyunsaturated fatty acids (PUFAs) in experimental stroke, leading to a better functional outcome. To assess the effects of administration of marine-derived n-3 PUFAs on functional outcomes and dependence in people with stroke.Our secondary outcomes were vascular-related death, recurrent events, incidence of other type of stroke, adverse events, quality of life, and mood. We searched the Cochrane Stroke Group trials register (6 August 2018), the Cochrane Central Register of Controlled Trials (CENTRAL; Issue 1, January 2019), MEDLINE Ovid (from 1948 to 6 August 2018), Embase Ovid (from 1980 to 6 August 2018), CINAHL EBSCO (Cumulative Index to Nursing and Allied Health Literature; from 1982 to 6 August 2018), Science Citation Index Expanded ‒ Web of Science (SCI-EXPANDED), Conference Proceedings Citation Index-Science - Web of Science (CPCI-S), and BIOSIS Citation Index. We also searched ongoing trial registers, reference lists, relevant systematic reviews, and used the Science Citation Index Reference Search. We included randomised controlled trials (RCTs) comparing marine-derived n-3 PUFAs to placebo or open control (no placebo) in people with a history of stroke or transient ischaemic attack (TIA), or both. At least two review authors independently selected trials for inclusion, extracted data, assessed risk of bias, and used the GRADE approach to assess the quality of the body of evidence. We contacted study authors for clarification and additional information on stroke/TIA participants. We conducted random-effects meta-analysis or narrative synthesis, as appropriate. The primary outcome was efficacy (functional outcome) assessed using a validated scale e.g. Glasgow Outcome Scale Extended (GOSE) dichotomised into poor or good clinical outcome, Barthel Index (higher score is better; scale from 0 to 100) or Rivermead Mobility Index (higher score is better; scale from 0 to 15). We included 29 RCTs; nine of them provided outcome data (3339 participants). Only one study included participants in the acute phase of stroke (haemorrhagic). Doses of marine-derived n-3 PUFAs ranged from 400 mg/day to 3300 mg/day. Risk of bias was generally low or unclear in most trials, with a higher risk of bias in smaller studies. We assessed results separately for short (up to three months) and longer (more than three months) follow-up studies.Short follow-up (up to three months)Functional outcome was reported in only one pilot study as poor clinical outcome assessed with GOSE (risk ratio (RR) 0.78, 95% confidence interval (CI) 0.36 to 1.68; 40 participants; very low quality evidence). Mood (assessed with GHQ-30, lower score better), was reported by only one study and favoured control (mean difference (MD) 1.41, 95% CI 0.07 to 2.75; 102 participants; low-quality evidence).We found no evidence of an effect of the intervention for the remainder of the secondary outcomes: vascular-related death (two studies, not pooled due to differences in population, RR 0.33, 95% CI 0.01 to 8.00, and RR 0.33, 95% CI 0.01 to 7.72; 142 participants; low-quality evidence); recurrent events (RR 0.41, 95% CI 0.02 to 8.84; 18 participants; very low quality evidence); incidence of other type of stroke (two studies, not pooled due to different type of index stroke, RR 6.11, 95% CI 0.33 to 111.71, and RR 0.63, 95% CI 0.25 to 1.58; 58 participants; very low quality evidence); and quality of life (physical component mean difference (MD) -2.31, 95% CI -4.81 to 0.19, and mental component MD -2.16, 95% CI -5.91 to 1.59; one study; 102 participants; low-quality evidence).Adverse events were reported by two studies (57 participants; very low quality evidence), one trial reporting extracranial haemorrhage (RR 0.25, 95% CI 0.04 to 1.73) and the other one reporting bleeding complications (RR 0.32, 95% CI 0.01 to 7.35).Longer follow-up (more than three months)One small trial assessed functional outcome with both Barthel Index (MD 7.09, 95% CI -5.16 to 19.34) for activities of daily living, and Rivermead Mobility Index (MD 1.30, 95% CI -1.31 to 3.91) for mobility (52 participants; very low quality evidence). We carried out meta-analysis for vascular-related death (RR 1.02, 95% CI 0.78 to 1.35; five studies; 2237 participants; low-quality evidence) and fatal recurrent events (RR 0.69, 95% CI 0.31 to 1.55; three studies; 1819 participants; low-quality evidence).We found no evidence of an effect of the intervention for mood (MD 1.00, 95% CI -2.07 to 4.07; one study; 14 participants; low-quality evidence). Incidence of other type of stroke and quality of life were not reported.Adverse events (all combined) were reported by only one study (RR 0.94, 95% CI 0.56 to 1.58; 1455 participants; low-quality evidence). We are very uncertain of the effect of marine-derived n-3 PUFAs therapy on functional outcomes and dependence after stroke as there is insufficient high-quality evidence. More well-designed RCTs are needed, specifically in acute stroke, to determine the efficacy and safety of the intervention.Studies assessing functionality might consider starting the intervention as early as possible after the event, as well as using standardised clinically-relevant measures for functional outcomes, such as the modified Rankin Scale. Optimal doses remain to be determined; delivery forms (type of lipid carriers) and mode of administration (ingestion or injection) also need further consideration.