Merck
  • Biocompatibility and cellular compatibility of decellularized tracheal matrix derived from rabbits.
Biocompatibility and cellular compatibility of decellularized tracheal matrix derived from rabbits.
The International journal of artificial organs (2019-05-14)
Fangbiao Zhang, Zhihao Wang, Chunhui Zheng, Chun Zhao, Hongcan Shi, Shu Pan, Weidong Zhang
ZUSAMMENFASSUNG
To study the different concentrations of Triton X-100 and nuclease needed to remove cells from the tracheal matrix of rabbits and analyse their biocompatibility and cellular compatibility. Fifty tracheas were harvested from donor New Zealand rabbits. Thirty tracheas were randomly divided into five groups (n = 6 each). The tracheas in group A were untreated and served as a control group, and those in groups B, C, D and E were treated with different concentrations of Triton X-100 (1%, 2%, 3% and 4%), respectively. The tracheas of the five groups were assessed by histological observation, scanning electron microscopy and mechanical evaluation. The remaining 20 donor tracheas, which were divided into a control group and an optimally decellularized group, were used for xenogeneic transplantation and cell seeding. Many epithelial cells and cartilage cells were observed in the tracheas of group A. There were fewer cartilage cells in the tracheas of groups C, D and E than in the tracheas of groups A and B under histological observation. In scanning electron microscopy, there were many ciliated epithelial cells in the tracheas of group A; in groups B and C, the ciliated epithelial cells disappeared, but the basement membrane was intact. The basement membranes were broken in the tracheas of groups D and E. Implanted decellularized tracheas showed good biocompatibility. Bone marrow mesenchymal stem cells grown in the decellularized tracheal matrix grew well. Decellularized tracheal matrix obtained from rabbits by 2% Triton X-100 may be suitable for the construction of tissue-engineered trachea because of its favourable morphological and biomechanical properties as well as its biocompatibility and cellular compatibly.