Merck
  • Extracellular matrix stiffness regulates human airway smooth muscle contraction by altering the cell-cell coupling.
Extracellular matrix stiffness regulates human airway smooth muscle contraction by altering the cell-cell coupling.
Scientific reports (2019-07-04)
Samuel R Polio, Suzanne E Stasiak, Ryan R Jamieson, Jenna L Balestrini, Ramaswamy Krishnan, Harikrishnan Parameswaran
ZUSAMMENFASSUNG
For an airway or a blood vessel to narrow, there must be a connected path that links the smooth muscle (SM) cells with each other, and transmits forces around the organ, causing it to constrict. Currently, we know very little about the mechanisms that regulate force transmission pathways in a multicellular SM ensemble. Here, we used extracellular matrix (ECM) micropatterning to study force transmission in a two-cell ensemble of SM cells. Using the two-SM cell ensemble, we demonstrate (a) that ECM stiffness acts as a switch that regulates whether SM force is transmitted through the ECM or through cell-cell connections. (b) Fluorescent imaging for adherens junctions and focal adhesions show the progressive loss of cell-cell borders and the appearance of focal adhesions with the increase in ECM stiffness (confirming our mechanical measurements). (c) At the same ECM stiffness, we show that the presence of a cell-cell border substantially decreases the overall contractility of the SM cell ensemble. Our results demonstrate that connectivity among SM cells is a critical factor to consider in the development of diseases such as asthma and hypertension.