Merck
  • The non-steroidal anti-inflammatory drug diclofenac sodium induces abnormal embryogenesis and delayed lethal effects in early life stage zebrafish (Danio rerio).
The non-steroidal anti-inflammatory drug diclofenac sodium induces abnormal embryogenesis and delayed lethal effects in early life stage zebrafish (Danio rerio).
Journal of applied toxicology : JAT (2018-11-22)
Yoshifumi Horie, Takahiro Yamagishi, Ayano Yagi, Yoko Shintaku, Taisen Iguchi, Norihisa Tatarazako
ZUSAMMENFASSUNG
Diclofenac sodium, a non-steroidal anti-inflammatory drug widely used in both human and veterinary medicine, has been detected in aquatic environments; therefore, its ecotoxicological effects on aquatic organisms need to be clarified. Recently, toxicity testing using zebrafish (Danio rerio) embryos has been recommended from the point of view of animal welfare; therefore, we investigated the suitability of using sub-lethal endpoints observed during embryogenesis for predicting lethal effects in early life stage zebrafish exposed to diclofenac sodium. After exposure to diclofenac sodium (0.4-7.0 mg/L) from 2 hours post-fertilization to 30 days post-hatching, abnormal embryogenesis, characterized by the presence of edema and body curvature, was observed in the 7.0 mg/L exposure group but not in any other groups including controls. The body curvature was found to be the result of abnormal development of the spine. All abnormal embryos hatched without delay, but died within 1 week after hatching, suggesting that the combination of the sub-lethal endpoints of edema and abnormal development of the spine during embryogenesis may predict lethal effects in early life stage zebrafish exposed to diclofenac sodium. Further investigations to verify these findings are needed. The value of the no observed effect concentrations for the embryogenesis, survival and growth endpoints were 3.5, 1.8 and >3.5 mg/L, respectively.