Merck
  • Tristetraprolin-mediated hexokinase 2 expression regulation contributes to glycolysis in cancer cells.
Tristetraprolin-mediated hexokinase 2 expression regulation contributes to glycolysis in cancer cells.
Molecular biology of the cell (2019-01-17)
Dong Jun Kim, Mai-Tram Vo, Seong Hee Choi, Ji-Heon Lee, So Yeon Jeong, Chung Hwan Hong, Jong Soo Kim, Unn Hwa Lee, Hyung-Min Chung, Byung Ju Lee, Wha Ja Cho, Jeong Woo Park
ZUSAMMENFASSUNG
Hexokinase 2 (HK2) catalyzes the first step of glycolysis and is up-regulated in cancer cells. The mechanism has not been fully elucidated. Tristetraprolin (TTP) is an AU-rich element (ARE)-binding protein that inhibits the expression of ARE-containing genes by enhancing mRNA degradation. TTP expression is down-regulated in cancer cells. We demonstrated that TTP is critical for down-regulation of HK2 expression in cancer cells. HK2 mRNA contains an ARE within its 3'-UTR. TTP binds to HK2 3'-UTR and enhances degradation of HK2 mRNA. TTP overexpression decreased HK2 expression and suppressed the glycolytic capacity of cancer cells, measured as glucose uptake and production of glucose-6-phosphate, pyruvate, and lactate. TTP overexpression reduced both the extracellular acidification rate (ECAR) and the oxygen consumption rate (OCR) of cancer cells. Ectopic expression of HK2 in cancer cells attenuated the reduction in glycolytic capacity, ECAR, and OCR from TTP. Taken together, these findings suggest that TTP acts as a negative regulator of HK2 expression and glucose metabolism in cancer cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%