Merck
  • Injury to hypothalamic Sim1 neurons is a common feature of obesity by exposure to high-fat diet in male and female mice.
Injury to hypothalamic Sim1 neurons is a common feature of obesity by exposure to high-fat diet in male and female mice.
Journal of neurochemistry (2019-01-08)
Eugene Nyamugenda, Marcus Trentzsch, Susan Russell, Tiffany Miles, Gunnar Boysen, Kevin D Phelan, Giulia Baldini
ZUSAMMENFASSUNG
The hypothalamus is essential for regulation of energy homeostasis and metabolism. Feeding hypercaloric, high-fat (HF) diet induces hypothalamic arcuate nucleus injury and alters metabolism more severely in male than in female mice. The site(s) and extent of hypothalamic injury in male and female mice are not completely understood. In the paraventricular nucleus (PVN) of the hypothalamus, single-minded family basic helix-loop helix transcription factor 1 (Sim1) neurons are essential to control energy homeostasis. We tested the hypothesis that exposure to HF diet induces injury to Sim1 neurons in the PVN of male and female mice. Mice expressing membrane-bound enhanced green fluorescent protein (mEGFP) in Sim1 neurons (Sim1-Cre:Rosa-mEGFP mice) were generated to visualize the effects of exposure to HF diet on these neurons. Male and female Sim1-Cre:Rosa-mEGFP mice exposed to HF diet had increased weight, hyperleptinemia, and developed hepatosteatosis. In male and female mice exposed to HF diet, expression of mEGFP was reduced by > 40% in Sim1 neurons of the PVN, an effect paralleled by cell apoptosis and neuronal loss, but not by microgliosis. In the arcuate nucleus of the Sim1-Cre:Rosa-mEGFP male mice, there was decreased alpha-melanocyte-stimulating hormone in proopiomelanocortin neurons projecting to the PVN, with increased cell apoptosis, neuronal loss, and microgliosis. These defects were undetectable in the arcuate nucleus of female mice exposed to the HF diet. Thus, injury to Sim1 neurons of the PVN is a shared feature of exposure to HF diet in mice of both sexes, while injury to proopiomelanocortin neurons in arcuate nucleus is specific to male mice. OPEN SCIENCE BADGES: This article has received a badge for *Open Materials* because it provided all relevant information to reproduce the study in the manuscript. The complete Open Science Disclosure form for this article can be found at the end of the article. More information about the Open Practices badges can be found at https://cos.io/our-services/open-science-badges/.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%