Merck
  • Purification and characterization of exo-β-1,4-glucosaminidase produced by chitosan-degrading fungus, Penicillium sp. IB-37-2A.
Purification and characterization of exo-β-1,4-glucosaminidase produced by chitosan-degrading fungus, Penicillium sp. IB-37-2A.
World journal of microbiology & biotechnology (2019-01-19)
Gleb E Aktuganov, Nailya F Galimzianova, Elena A Gilvanova, Elena A Pudova, Lyudmila Yu Kuzmina, Alexander I Melentiev, Violetta R Safina
ZUSAMMENFASSUNG
Chitosan-degrading fungal strain, Penicillium sp. IB-37-2A, produced mainly extracellular chitosanolytic enzymes under submerged agitating cultivation in presence of soluble chitosan or colloidal chitin as main carbon source. Significant N-acetyl-β-D-glucosaminidase activity (8-18 × 103 U·ml-1) was also detected in culture filtrate of the fungal strain. Alone major exo-chitosanase from culture filtrate of Penicillium sp. IB-37-2A was purified in 46-fold using ultrafiltration, affinity sorption on colloidal chitosan and hydrophobic chromatography on Phenyl-Sepharose CL 4B and characterized. Molecular weight of the exo-β-1.4-glucosaminidase is 41 kDa according to SDS-PAGE. The purified enzyme has optima pH and temperature 4.0 and 50-55 °C, respectively, pI 4.9; it is stable under pH 3.0-8.0 and 55 °C. Activity of the enzyme is strongly inhibited by 1 mM Hg2+ and Ag+, in less degree-10 mM Cu2+, Zn2+, Ni+ and Fe2+, slightly activated-with 1 mM Mg2+, 10 mM Ca2+, tween-80 (10 mM) and Triton X-100 (1 mM). Viscosimetric assay confirmed reported earlier exo-splitting manner of the enzyme activity. Soluble chitosan (deacetylation degree (DD) 80-85%) is most rapidly hydrolyzed by the enzyme (Vmax = 7.635 µM × min-1 × mg-1, KM ~ 0.83 mg/ml). Purified exo-chitosanase also degraded laminarin, β-glucan, colloidal chitin and showed significant chitobiohydrolase activity (V ~ 50 µM × ml-1 × min-1 for pNP-GlcNAc2) but no hydrolyzed CMC, cellulose, xylan and galactomannan. It is found that crude and partially purified exo-β-1.4-glucosaminidase inhibits in vitro the growth of some phytopathogenic fungi that is first report for antifungal activity of exo-chitosanase.