Merck
  • Long intergenic non-protein coding RNA 511 promotes the progression of osteosarcoma cells through sponging microRNA 618 to upregulate the expression of maelstrom.
Long intergenic non-protein coding RNA 511 promotes the progression of osteosarcoma cells through sponging microRNA 618 to upregulate the expression of maelstrom.
Aging (2019-08-07)
Wen Guo, Qing Yu, Ming Zhang, Fang Li, Yu Liu, Weiwei Jiang, Haitao Jiang, Haijun Li
ZUSAMMENFASSUNG
Osteosarcoma is a tumor disease that commonly exists among young populations. Our research explored the role of the LINC00511/microRNA-618/MAEL axis in osteosarcoma.Expression profiles of long non-coding RNAs (lncRNAs) in osteosarcoma (OS) tissues were constructed, and LINC00511 expression levels were verified with qRT-PCR. LncRNA-miRNA and miRNA-mRNA interactions were predicted. Validation was performed using a dual-luciferase reporter assay. Protein expression levels of MAEL were evaluated by Western blot assays. The effects of LINC00511, miR-618 and MAEL on the proliferation, viability, and metastasis of OS cells were detected using colony formation, cell counting kit-8 (CCK-8) and transwell assays, respectively. The apoptosis rates of OS cells were investigated using flow cytometry. The tumor-suppressing effect of LINC00511 silencing was also analyzed using a xenograft model in nude mice.LINC00511 overexpression was observed in OS tissues and cell lines. Knockdown of LINC00511 in nude mice inhibited the tumorigenic ability of OS cells. Transfection-induced overexpression of LINC00511 and MAEL, as well as downregulation, highlighted the features of tumor cells, and LINC00511 overexpression reduced apoptosis in vitro.LINC00511 was confirmed to be beneficial for osteosarcoma development via sponging miR-618 and increasing MAEL expression and may thus be considered a potential target for osteosarcoma therapy.