Merck
The anticonvulsive Phenhydan® suppresses extrinsic cell death.
Cell death and differentiation (2018-11-18)
Caroline Moerke, Isabel Jaco, Christin Dewitz, Tammo Müller, Annette V Jacobsen, Jérémie Gautheron, Jürgen Fritsch, Jessica Schmitz, Jan Hinrich Bräsen, Claudia Günther, James M Murphy, Ulrich Kunzendorf, Pascal Meier, Stefan Krautwald
ZUSAMMENFASSUNG
Different forms of regulated cell death-like apoptosis and necroptosis contribute to the pathophysiology of clinical conditions including ischemia-reperfusion injury, myocardial infarction, sepsis, and multiple sclerosis. In particular, the kinase activity of the receptor-interacting serine/threonine protein kinase 1 (RIPK1) is crucial for cell fate in inflammation and cell death. However, despite its involvement in pathological conditions, no pharmacologic inhibitor of RIPK1-mediated cell death is currently in clinical use. Herein, we screened a collection of clinical compounds to assess their ability to modulate RIPK1-mediated cell death. Our small-scale screen identified the anti-epilepsy drug Phenhydan® as a potent inhibitor of death receptor-induced necroptosis and apoptosis. Accordingly, Phenhydan® blocked activation of necrosome formation/activation as well as death receptor-induced NF-κB signaling by influencing the membrane function of cells, such as lipid raft formation, thus exerting an inhibitory effect on pathophysiologic cell death processes. By targeting death receptor signaling, the already FDA-approved Phenhydan® may provide new therapeutic strategies for inflammation-driven diseases caused by aberrant cell death.