Merck
  • Reducing the risk of post-endoscopic retrograde cholangiopancreatography pancreatitis using 4-Fr pancreatic plastic stents placed with common-type guidewires: Results from a prospective multinational registry.
Reducing the risk of post-endoscopic retrograde cholangiopancreatography pancreatitis using 4-Fr pancreatic plastic stents placed with common-type guidewires: Results from a prospective multinational registry.
Digestive endoscopy : official journal of the Japan Gastroenterological Endoscopy Society (2018-12-07)
Nadav Sahar, Andrew Ross, Sundeep Lakhtakia, Gregory A Coté, Horst Neuhaus, Marco J Bruno, Oleh Haluszka, Richard Kozarek, Mohan Ramchandani, Torsten Beyna, Jan W Poley, Jennifer Maranki, Martin Freeman, Prashant Kedia, Paul Tarnasky
ZUSAMMENFASSUNG
Pancreatic plastic stents (PPS) can reduce the risk of post-endoscopic retrograde cholangiopancreatography pancreatitis (PEP). Prospective multicenter documentation of PEP rate after PPS placement is scarce. A new 4-Fr stent designed to be deployed over a 0.035-inch guidewire was used to assess the effectiveness of PEP prophylaxis. High-PEP-risk patients received a 4-Fr PPS for primary or secondary prophylaxis at seven centers in four countries. Patients were followed until spontaneous PPS migration, endoscopic stent removal, or for 4 months, whichever came first. Main outcome was PEP rate. One hundred six (106) patients received PPS for PEP prophylaxis [61 (58%) primary, 45 (42%) secondary prophylaxis]. Median age was 54 years. Eighty-one (76%) PPS were placed using a 0.035-inch guidewire. By investigator choice 99 (93%) stents were single pigtail. Median stent length was 8 cm (range 3-12 cm). Technical success achieved in 100% of cases. Two patients in the primary prophylaxis group (3%, 95% CI 0.4-11%) experienced mild/moderate PEP. Seventy-eight PPS available for analysis underwent spontaneous migration after a median of 29 days. There were no reports of stent-induced ductal trauma. Post-hoc analysis of migration rate by PPS length showed no statistically significant trend. Among high-risk patients in the primary prophylaxis group, observed rates of PEP are low (3%, 95% CI 0.4-11%) with the use of prophylactic 4-Fr pancreatic duct stents compatible with a 0.035-inch guidewire. This low rate is not unequivocally due to the prophylactic stent.