Merck
  • Nearly half of Ultrio plus NAT non-discriminated reactive blood donors were identified as occult HBV infection in South China.
Nearly half of Ultrio plus NAT non-discriminated reactive blood donors were identified as occult HBV infection in South China.
BMC infectious diseases (2019-07-05)
Xianlin Ye, Tong Li, Wen Shao, Jinfeng Zeng, Wenxu Hong, Liang Lu, Weigang Zhu, Chengyao Li, Tingting Li
ZUSAMMENFASSUNG
Blood donor plasma samples were detected by the Ultrio Plus NAT system for HBV, HCV and HIV-1 in Shenzhen blood center, China. Reactive samples underwent further discriminatory testing of a single virus by the same methodology. A large number of cases of non-discriminated reactive (NDR) donors were found, leaving potential risk of transmitting HBV if not deferrals. This study identified those non-discriminated samples. The NDR plasma samples from blood donation screening were detected and classified by additional molecular and serological tests. Molecular characterizations of DNA+ NDR were determined by sequencing analysis. A number of 259 (0.21%) NDR plasma samples from screening of 123,280 eligible blood donors were detected, which presented a higher rate (91.1%) of anti-HBc reactivity and nearly half (46.7%) of HBV DNA+ that classified as occult HBV infection (OBI). Most OBI strains were wild-type HBV, but some substitutions V168A, S174 N, V177A, Q129R/L/H, G145A/R in S region of genotype B (OBIB) and T47K/V/A, P49H/L, Q101R/H/K, S174 N, L175S, V177A, T118 M/R/K, G145R/A/K/E, R160K/N in S region of genotype C (OBIC) strains were identified in high frequency. Nearly half of NDR blood samples were identified as OBI, in which a number of important mutations were detected. NDR donation might have potential risk for HBV transmission, but need to be further investigated.