Merck
  • Occurrence, Distribution, and Potential Sources of Organophosphate Esters in Urban and Rural Surface Water in Shanghai, China.
Occurrence, Distribution, and Potential Sources of Organophosphate Esters in Urban and Rural Surface Water in Shanghai, China.
Archives of environmental contamination and toxicology (2019-05-28)
Zhenyong Zhang, Haiyang Shao, Minghong Wu, Junyun Zhang, Dongyang Li, Jinsong Li, Hongyong Wang, Wenyan Shi, Gang Xu
ZUSAMMENFASSUNG
In this study, the occurrence and distribution patterns of eight organophosphate esters (OPEs) were investigated in urban and rural surface water in a typical cosmopolitan city: Shanghai, China. In addition, concentration levels and removal efficiencies of seven sewage treatment plants were analyzed. The OPEs concentrations detected in urban rivers were significantly higher than those detected in rural rivers. Total OPEs ranged from 185.4 to 321 ng L-1 in rural surface water and from 340 to 1688.7 ng L-1 in urban, with an average of 221.8 ng L-1 and 850.2 ng L-1, respectively. Compared with other studies published in the world, the OPEs contamination in surface river water in Shanghai was at a moderate level. Furthermore, the potential sources of OPEs in urban surface water were investigated, and the results indicated that OPEs in urban surface water mainly came from three potential sources. In rural surface water, the OPE concentrations were uniformly distributed, so OPEs in rural surface water may came from nonpoint source pollution. Last, a preliminary environmental risk assessment and health risk assessment were conducted. The results showed low environmental risks at all sampling sites (except for sampling point R7: medium risk for algae) for the three aquatic organisms (algae, daphnia, and fish). Health risk assessment indicated a noncarcinogenic risk for diverse human groups for ƩOPEs.