Merck
  • Thermal Decomposition Kinetics and Mechanism of In-Situ Prepared Bio-based Poly(propylene 2,5-furan dicarboxylate)/Graphene Nanocomposites.
Thermal Decomposition Kinetics and Mechanism of In-Situ Prepared Bio-based Poly(propylene 2,5-furan dicarboxylate)/Graphene Nanocomposites.
Molecules (Basel, Switzerland) (2019-05-06)
Zoi Terzopoulou, Evangelia Tarani, Nejib Kasmi, Lazaros Papadopoulos, Konstantinos Chrissafis, Dimitrios G Papageorgiou, George Z Papageorgiou, Dimitrios N Bikiaris
ZUSAMMENFASSUNG
Bio-based polyesters are a new class of materials that are expected to replace their fossil-based homologues in the near future. In this work, poly(propylene 2,5-furandicarboxylate) (PPF) nanocomposites with graphene nanoplatelets were prepared via the in-situ melt polycondensation method. The chemical structure of the resulting polymers was confirmed by 1H-NMR spectroscopy. Thermal stability, decomposition kinetics and the decomposition mechanism of the PPF nanocomposites were studied in detail. According to thermogravimetric analysis results, graphene nanoplatelets did nοt affect the thermal stability of PPF at levels of 0.5, 1.0 and 2.5 wt.%, but caused a slight increase in the activation energy values. Pyrolysis combined with gas chromatography and mass spectroscopy revealed that the decomposition mechanism of the polymer was not altered by the presence of graphene nanoplatelets but the extent of secondary homolytic degradation reactions was increased.