Merck
  • MIF 173 G>C variation was associated with depressive disorder in type 2 diabetes in an Iranian population.
MIF 173 G>C variation was associated with depressive disorder in type 2 diabetes in an Iranian population.
Psychoneuroendocrinology (2019-03-25)
Armita Kakavand Hamidi, Seyed Masoud Arzaghi, Mostafa Qorbani, Fatemeh Khatami, Mehdi Ebrahimi, Fatemeh Bandarian, Samaneh Enayati, Mahsa M Amoli
ZUSAMMENFASSUNG
Type 2 diabetes mellitus (T2DM) is a continuous metabolic disease linked with increased rate of mortality and morbidity. High levels of glucose can damage organs including kidneys, eyes, and the nervous system. Individuals with T2DM have a high prevalence of major depression. One possible question we aimed to address was the extent of co-occurrence of diabetes and depression resulting from correlated genetic risk factors. The current study aimed to investigate the possible associations between the macrophage migration inhibitory factor (MIF) functional variant and the risk of developing depression in T2DM patients. The study groups consisted of 120 patients with T2DM and comorbid depression and 120 patients with T2DM, without depression, who were recruited from the same region. Genotyping of the MIF -173 G > C (rs755622) variant was performed using Polymerase Chain Reaction (PCR) and Restriction Fragment Length Polymorphism (RFLP). In addition, the level of MIF expression was comparatively evaluated in both groups by quantitative real-time PCR. The data showed that the presence of C allele (GC + CC vs. GG) might predispose females to depression in patients with T2DM. In addition, patients with T2DM carrying at least one C allele showed significantly elevated levels of MIF RNA expression in comparison to individuals with GG genotype. MIF variant could be considered as a factor making female patients with T2DM vulnerable to depression. So, this might be an important result for precise diagnosis and/or earlier treatment.