Merck
  • Increasing importance of European lineages in seeding the hepatitis C virus subtype 1a epidemic in Spain.
Increasing importance of European lineages in seeding the hepatitis C virus subtype 1a epidemic in Spain.
Euro surveillance : bulletin Europeen sur les maladies transmissibles = European communicable disease bulletin (2019-03-14)
Ana Belen Pérez, Bram Vrancken, Natalia Chueca, Antonio Aguilera, Gabriel Reina, Miguel García-Del Toro, Francisco Vera, Miguel Angel Von Wichman, Juan Ignacio Arenas, Francisco Téllez, Juan A Pineda, Mohamed Omar, Enrique Bernal, Antonio Rivero-Juárez, Elisa Fernández-Fuertes, Alberto de la Iglesia, Juan Manuel Pascasio, Philippe Lemey, Féderico Garcia, Lize Cuypers
ZUSAMMENFASSUNG
BackgroundReducing the burden of the hepatitis C virus (HCV) requires large-scale deployment of intervention programmes, which can be informed by the dynamic pattern of HCV spread. In Spain, ongoing transmission of HCV is mostly fuelled by people who inject drugs (PWID) infected with subtype 1a (HCV1a).AimOur aim was to map how infections spread within and between populations, which could help formulate more effective intervention programmes to halt the HCV1a epidemic in Spain.MethodsEpidemiological links between HCV1a viruses from a convenience sample of 283 patients in Spain, mostly PWID, collected between 2014 and 2016, and 1,317, 1,291 and 1,009 samples collected abroad between 1989 and 2016 were reconstructed using sequences covering the NS3, NS5A and NS5B genes. To efficiently do so, fast maximum likelihood-based tree estimation was coupled to a flexible Bayesian discrete phylogeographic inference method.ResultsThe transmission network structure of the Spanish HCV1a epidemic was shaped by continuous seeding of HCV1a into Spain, almost exclusively from North America and European countries. The latter became increasingly relevant and have dominated in recent times. Export from Spain to other countries in Europe was also strongly supported, although Spain was a net sink for European HCV1a lineages. Spatial reconstructions showed that the epidemic in Spain is diffuse, without large, dominant within-country networks.ConclusionTo boost the effectiveness of local intervention efforts, concerted supra-national strategies to control HCV1a transmission are needed, with a strong focus on the most important drivers of ongoing transmission, i.e. PWID and other high-risk populations.