Merck
  • C1q/TNF-related protein-9 promotes macrophage polarization and improves cardiac dysfunction after myocardial infarction.
C1q/TNF-related protein-9 promotes macrophage polarization and improves cardiac dysfunction after myocardial infarction.
Journal of cellular physiology (2019-04-07)
Mingxin Liu, Lin Yin, Wei Li, Juan Hu, Huibo Wang, Bingjie Ye, Yanhong Tang, Congxin Huang
ZUSAMMENFASSUNG
The timely regulation of inflammatory M1 macrophage polarization toward regenerative M2 macrophages suggests the possibility of immunotherapy after myocardial infarction (MI). C1q/TNF-related protein-9 (CTRP9) has anti-inflammatory effects and can ameliorate heart function in mice after long-term myocardial infarction. The role of CTRP9 in macrophage polarization remains completely unclear. This study determined whether CTRP9 can preserve post-MI early cardiac function through the regulation of macrophage polarization. In the present study, an adenovirus-delivered CTRP9 supplement promoted macrophage polarization at Day 3 post MI and improved cardiac function at Day 7 post MI. Pretreatment with gCTRP9 promoted the M1 to M2 polarization transition and attenuated inflammation after lipopolysaccharide + interferon-γ stimulation; the effects were partly abrogated by the adenosine monophosphate kinase (AMPK) inhibitor compound C and were obviously reinforced by pyrrolidine dithiocarbamate, a nuclear factor-κB (NF-κB) inhibitor. Meanwhile, CTPR9 markedly reduced the expression of toll-like receptor 4 (TLR4), myeloid differentiation factor 88 (MyD88), and NF-κB p65 phosphorylation by promoting AMPK phosphorylation in vivo and in vitro. Moreover, the competitive binding of gCTRP9 and LPS to the myeloid differentiation protein 2 (MD2)/TLR4 complex was associated with direct binding to MD2, thereby inhibiting the downstream signaling molecule MyD88. Taken together, we demonstrated that CTRP9 improved post-MI early cardiac function, at least in part, by modulating M1/M2 macrophage polarization, largely via the TLR4/MD2/MyD88 and AMPK-NF-κB pathways.