Merck
Study of Myocardial Perfusion in Obese Individuals without Known Ischemic Heart Disease.
Arquivos brasileiros de cardiologia (2018-12-21)
Tufi Dippe, Cláudio Leinig Pereira da Cunha, Rodrigo Julio Cerci, Arnaldo Lafitte Stier, João Vicente Vítola
ZUSAMMENFASSUNG
Obesity is associated with an increased risk of type 2 diabetes mellitus (DM), ischemic heart disease (IHD) and cardiovascular mortality. Several studies have demonstrated the diagnostic and prognostic value of single photon computed tomography-myocardial perfusion scintigraphy (SPECT-MPI) in the evaluation of patients with suspected IHD, including in obese population. Data on clinical risk factors and their association with abnormal myocardial perfusion in obese patients are scarce in the Brazilian population. To determine the factors associated with abnormal myocardial perfusion in obese individuals without known IHD. We studied obese patients without known IHD who were referred for evaluation through SPECT-MPI between January 2011 and December 2016. Clinical variables and results of SPECT-MPI were obtained systematically. The distribution of continuous variables was assessed using the Shapiro-Wilk and Shapiro-Francia tests. We used the unpaired Student t test to compare the means of continuous variables with normal distribution and the Chi Square test for binomial variables analysis. A p value < 0.05 was considered statistically significant. The association of the clinical variables for the presence of factors associated with abnormal myocardial perfusion was determined by univariate and multivariate logistic regression analysis, and respective odds ratios (OR) and 95% confidence intervals (CI). The study sample consisted of 5,526 obese patients. Mean body mass index (BMI) of our patients was 33.9 ± 3.7 kg/m2, 31% had DM, and myocardial perfusion abnormalities was observed in 23% of the total sample. The factors associated with abnormal myocardial perfusion on multivariate analysis were: age (OR: 1.02, 95% CI 1.01-1.03, p < 0.001), DM (OR: 1.57, 95% CI 1.31-1.88, p < 0.001), typical angina before the test (OR: 2.45, 95% CI: 1.82-3.31, p < 0.001), need for pharmacologic stress test (OR: 1.61, 95% CI: 1.26-2.07, p < 0.001), less physical effort evaluated in metabolic equivalents (METs) during the exercise treadmill test (OR: 0.89, 95% CI: 0.85-0.94, p < 0.001) and a lower post-stress left ventricular ejection fraction after stress (LVEF; OR: 0.989, 95% CI: 0.984-0.994, p < 0.001). The factors associated with abnormal myocardial perfusion in obese patients without known IHD were age, DM, presence of typical angina, ventricular dysfunction, and inability to undergo physical stress as clinical variables, in addition to functional capacity during physical stress.