Merck
  • Design and Synthesis of a Cyclic Double-Grafted Polymer Using Active Ester Chemistry and Click Chemistry via A "Grafting onto" Method.
Design and Synthesis of a Cyclic Double-Grafted Polymer Using Active Ester Chemistry and Click Chemistry via A "Grafting onto" Method.
Polymers (2019-04-10)
Meng Liu, Lu Yin, Shuangshuang Zhang, Zhengbiao Zhang, Wei Zhang, Xiulin Zhu
ZUSAMMENFASSUNG
Combing active ester chemistry and click chemistry, a cyclic double-grafted polymer was successfully demonstrated via a "grafting onto" method. Using active ester chemistry as post-functionalized modification approach, cyclic backbone (c-P2) was synthesized by reacting propargyl amine with cyclic precursor (poly(pentafluorophenyl 4-vinylbenzoate), c-PPF4VB6.5k). Hydroxyl-containing polymer double-chain (l-PS-PhOH) was prepared by reacting azide-functionalized polystyrene (l-PSN₃) with 3,5-bis(propynyloxy)phenyl methanol, and further modified by azide group to generate azide-containing polymer double-chain (l-PS-PhN₃). The cyclic backbone (c-P2) was then coupled with azide-containing polymer double-chain (l-PS-PhN₃) via CuAAC reaction to construct a novel cyclic double-grafted polymer (c-P2-g-Ph-PS). This research realized diversity and complexity of side chains on cyclic-grafted polymers, and this cyclic double-grafted polymer (c-P2-g-Ph-PS) still exhibited narrow molecular weight distribution (Mw/Mn < 1.10).