Merck
  • Over-expression of miR-206 decreases the Euthyrox-resistance by targeting MAP4K3 in papillary thyroid carcinoma.
Over-expression of miR-206 decreases the Euthyrox-resistance by targeting MAP4K3 in papillary thyroid carcinoma.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2019-03-25)
Fangzhou Liu, Rong Yin, Xinyuan Chen, Wei Chen, Yichun Qian, Yanbin Zhao, Yuan Jiang, Dawei Ma, Tingting Hu, Tonghua Yu, Yan Zhu, Yuan Zhang
ZUSAMMENFASSUNG
microRNAs (miRNAs) play a critical role in drug resistance of multiple cancers including papillary thyroid carcinoma (PTC), indicating the potential of miRNAs as chemoresistance regulators in cancer treatment. The aim of this paper is to explore the relationship between miR-206 and chemoresistance of PTC. qRT-PCR was conducted to examine the expression of miR-206 in PTC tissues, parental and TPC-1/euthyrox. The CCK-8 assay, EdU assay and flow cytometry were performed to test cells viability, proliferation and apoptosis, respectively. Luciferase reporter assay was used to confirm the potential target of miR-206. Western blotting analysis was performed to evaluate the expressions of related-proteins. miR-206 was significantly down-regulated in PTC tissues, parental and TPC-1/euthyrox. Moreover, the expression of miR-206 was exceptionally lower in TPC-1/euthyrox cells than that in TPC-1 cells. Furthermore, we found that over-expression of miR-206 could notably decrease the IC50 values both in TPC-1 and TPC-1/euthyrox cells, which indicated that miR-206 played an essential role in the euthyrox resistance in PTC. In addition, up-regulation of miR-206 inhibited the proliferation, induced apoptosis, suppressed the expressions of multidrug resistance-related proteins, including p-gp, MRP, BCRP and LRP, in euthyrox-resistant PTC cells. Besides, over-expression of miR-206 could notably promoted the expression of NIS, an intrinsic membrane protein that mediates the active transport of iodide into the thyroid and other tissues, playing a critical role in the progress. Further, miR-206 was demonstrated to be able to bind to MAP4K3 and negatively regulated the expression of MAP4K3. Besides, MAP4K3 was clearly up-regulated in PTC tissues, parental and TPC-1/euthyrox cells, and down-regulation of miR-206 attenuated the effect of si-MAP4K3 on the euthyrox sensitivity in euthyrox-resistant PTC cells. Moreover, TPC-1/euthyrox cells transfected with miR-206 mimics could significantly inhibit the expressions of p-p38, p-JNK and p-Erk, which indicated that miR-206 might play an essential role in the euthyrox resistance in PTC by negatively regulating the p38 and JNK signaling pathway. miR-206 contributed to euthyrox resistance in PTC cells through blockage p38 and JNK signaling pathway by targeting MAP4K3, providing a potential therapeutic application for the treatment of patients with euthyrox-resistant PTC in the further.