Merck
  • Photocontrolled reversible supramolecular assemblies of an azobenzene-containing surfactant with α-cyclodextrin
Photocontrolled reversible supramolecular assemblies of an azobenzene-containing surfactant with α-cyclodextrin
Angewandte Chemie (International ed. in English) (2007)
Wang Y, et al.