- Novel IKK inhibitors: beta-carbolines.
Novel IKK inhibitors: beta-carbolines.
Bioorganic & medicinal chemistry letters (2003-06-26)
Alfredo C Castro, Luan C Dang, François Soucy, Louis Grenier, Hormoz Mazdiyasni, Maria Hottelet, Lana Parent, Christine Pien, Vito Palombella, Julian Adams
PMID12824047
ZUSAMMENFASSUNG
Inhibitors of IkappaB kinase (IKK) have long been sought as specific regulators of NF-kappaB. A screening effort of the endogenous IKK complex allowed us to identify 5-bromo-6-methoxy-beta-carboline as a nonspecific IKK inhibitor. Optimization of this beta-carboline natural product derivative resulted in a novel class of selective IKK inhibitors with IC(50)s in the nanomolar range. In addition, we show that one of these beta-carboline analogues inhibits the phosphorylation of IkappaBalpha and subsequent activation of NF-kappaB in whole cells, as well as blocking TNF-alpha release in LPS-challenged mice.