Merck
  • Both the cytosols and detergent extracts of breast cancer tissues are suited to evaluate the prognostic impact of the urokinase-type plasminogen activator and its inhibitor, plasminogen activator inhibitor type 1.
Both the cytosols and detergent extracts of breast cancer tissues are suited to evaluate the prognostic impact of the urokinase-type plasminogen activator and its inhibitor, plasminogen activator inhibitor type 1.
Cancer research (1994-05-15)
F Jänicke, L Pache, M Schmitt, K Ulm, C Thomssen, A Prechtl, H Graeff
ZUSAMMENFASSUNG
The serine protease urokinase-type plasminogen activator (uPA) plays a key role in tumor-associated proteolysis in malignant solid tumors. Proteolytic activity of uPA is controlled by its naturally occurring plasminogen activator inhibitor type 1. As an initial observation, a correlation of enzymatic uPA activity in breast cancer cytosols with prognosis was described in 1988 (Duffy et al., Cancer (Phila.), 62: 531-533, 1988). A pronounced prognostic impact of uPA, independent of classical risk parameters, was then first demonstrated in detergent-extracted (Triton X-100) breast cancer tissues by applying enzyme-linked immunosorbent assay techniques (Jänicke et al., Lancet, 2: 1049, 1989; Fibrinolysis, 4:69-78, 1990; Duffy et al., Cancer Res., 50: 6827-6829, 1990). In addition, not only uPA but also plasminogen activator inhibitor type 1 were shown to be of prognostic value in breast cancer (Jänicke et al., Semin. Thromb. Hemostasis, 17: 303-312, 1991; Breast Cancer Res. Treat., 24: 195-208, 1993). Subsequently, the prognostic value of uPA and plasminogen activator inhibitor type 1 was also confirmed in studies using archived "cytosol fractions" of breast cancer tissues (Foekens et al., Cancer Res., 52: 6101-6105, 1992; Spyratos et al., J. Natl. Cancer Inst., 84: 1266-1272, 1992; Grondahl-Hansen et al., Cancer Res., 53: 2513-2521, 1993; Sumiyoshi et al., Int. J. Cancer, 50: 345-348, 1992). A direct comparison of both methods with regard to prognosis, however, was lacking. We therefore prepared both the detergent-treated tissue extracts and the cytosol fractions from the same breast cancer specimens to allow a direct comparison of both methods. In 247 breast cancer patients investigated, the Triton X-100-extracted tissues revealed about twice as much uPA antigen (uPATx: median, 2.32 ng/mg protein) than the cytosol fractions (uPAcyt: median, 1.07 ng/mg protein). In contrast, the presence of Triton X-100 did not result in an increase of PAI-1 (PAI-1Tx: median, 6.34 ng PAI-1/mg protein) compared to the cytosol fractions (PAI-1cyt: median, 7.15 ng PAI-1/mg protein). Good correlations between uPATx and uPAcyt (R = 0.72) and between PAI-1Tx and PAI-1cyt (R = 0.88) were observed. Furthermore, PAI-1 and uPA are moderately correlated with each other (uPATx versus PAI-1Tx: R = 0.40; uPAcyt versus PAI-1cyt: R = 0.39). The prognostic power of uPA showed its best advantage in Triton X-100-extracted tissues (RR = 3.22), most pronounced in the subgroups of node-negative and premenopausal patients, respectively.(ABSTRACT TRUNCATED AT 400 WORDS)
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
uPA-Aktivitätsassay-Kit, The uPA Activity Assay Kit provides a quick, efficient & sensitive system for evaluation of uPA activity & for screening of uPA inhibitors.