Merck
  • Novel therapy for malignant pleural mesothelioma based on anti-energetic effect: an experimental study using 3-Bromopyruvate on nude mice.
Novel therapy for malignant pleural mesothelioma based on anti-energetic effect: an experimental study using 3-Bromopyruvate on nude mice.
Anticancer research (2009-05-06)
Xiaodong Zhang, Emilie Varin, Mélanie Briand, Stéphane Allouche, Natacha Heutte, Laurent Schwartz, Laurent Poulain, Philippe Icard
ZUSAMMENFASSUNG
Most cancer cells exhibit increased anaerobic glycolysis and use this metabolic pathway for the generation of ATP as a main source of energy. This impaired metabolism of glucose, leading to the secretion of lactic acid even in the presence of oxygen, is named the Warburg effect. Because cancer cells are partly or mainly dependent on such a pathway to generate ATP, inhibition of glycolysis may slow down the proliferation or kill cells. The effect of 3-Bromopyruvate (3-BrPA) alone or associated to cisplatin on nude mice presenting intraperitoneal carcinomatosis developed after intraperitoneal injection of human mesothelioma cells (MSTO-211H) was investigated. 3-BrPA significantly prolonged survival of animals. Combined with cisplatin, it demonstrated significant benefit on survival whereas cisplatin alone had no or a mild effect. 3-BrPA may thus constitute an interesting novel anticancer drug that could be tested in humans.