Merck
  • Determination of L-carnitine in food supplement formulations using ion-pair chromatography with indirect conductimetric detection
Determination of L-carnitine in food supplement formulations using ion-pair chromatography with indirect conductimetric detection
Journal of Chromatography A (2005)
Kakou A, et al.