Merck
  • Induction of DNA damage-inducible gene GADD45beta contributes to sorafenib-induced apoptosis in hepatocellular carcinoma cells.
Induction of DNA damage-inducible gene GADD45beta contributes to sorafenib-induced apoptosis in hepatocellular carcinoma cells.
Cancer research (2010-11-11)
Da-Liang Ou, Ying-Chun Shen, Sung-Liang Yu, Kuen-Feng Chen, Pei-Yen Yeh, Hsiang-Hsuan Fan, Wen-Chi Feng, Ching-Tzu Wang, Liang-In Lin, Chiun Hsu, Ann-Lii Cheng
ZUSAMMENFASSUNG
Markers that could accurately predict responses to the general kinase inhibitor sorafenib are needed to better leverage its clinical applications. In this study, we examined a hypothesized role in the drug response for the growth arrest DNA damage-inducible gene 45β (GADD45β), which is commonly underexpressed in hepatocellular carcinoma (HCC) where sorafenib may offer an important new therapeutic option. The anticancer activity of sorafenib-induced GADD45β expression was tested in a panel of HCC cell lines and xenograft models. We found that GADD45β mRNA and protein expression were induced relatively more prominently in HCC cells that were biologically sensitive to sorafenib treatment. GADD45β induction was not found after treatment with either the mitogen-activated protein kinase-extracellular signal-regulated kinase (ERK) kinase (MEK) inhibitor U0126 or the Raf inhibitor ZM336372, suggesting that GADD45β induction by sorafenib was independent of Raf/MEK/ERK signaling activity. However, c-Jun NH2-terminal kinase (JNK) kinase activation occurred preferentially in sorafenib-sensitive cells. Small interfering RNA-mediated knockdown of GADD45βor JNK kinase limited the proapoptotic effects of sorafenib in sorafenib-sensitive cells. We defined the -339/-267 region in the GADD45β promoter containing activator protein-1 and SP1-binding sites as a crucial region for GADD45β induction by sorafenib. Together, our findings suggest that GADD45β induction contributes to sorafenib-induced apoptosis in HCC cells, prompting further studies to validate its potential value in predicting sorafenib efficacy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
U0126, CAS-Nr. 109511-58-2, Calbiochem, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC50 = 72 nM) and MEK2 (IC50 = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.
Sigma-Aldrich
JNK-Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC50 = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).
Sigma-Aldrich
ZM 336372, A potent, cell-permeable, reversible, ATP-competitive and specific inhibitor of the protein kinase c-Raf (IC50 = 70 nM).