Merck
  • Functional properties and structural characterization of rice delta1-pyrroline-5-carboxylate reductase
Functional properties and structural characterization of rice delta1-pyrroline-5-carboxylate reductase
Frontiers in Plant Science (2015)
Forlani G, et al.