Merck
  • Anoxic metabolism and biochemical production in Pseudomonas putida F1 driven by a bioelectrochemical system
Anoxic metabolism and biochemical production in Pseudomonas putida F1 driven by a bioelectrochemical system
Biotechnology for Biofuels (2016)
Lai B, et al.