Merck
  • Expression and cellular localization of cyclooxygenases and prostaglandin E synthases in the hemorrhagic brain
Expression and cellular localization of cyclooxygenases and prostaglandin E synthases in the hemorrhagic brain
Journal of Neuroinflammation (2011)
Wu T, et al.