Merck
  • Space filling of beta-cyclodextrin and beta-cyclodextrin derivatives by volatile hydrophobic guests
Space filling of beta-cyclodextrin and beta-cyclodextrin derivatives by volatile hydrophobic guests
Beilstein Journal of Organic Chemistry (2013)
Fourmentin S, et al.