Merck
  • A new platform for chymotrypsin isolation from fresh bovine pancreas using an environmental friendly polyelectrolyte: Alginate
A new platform for chymotrypsin isolation from fresh bovine pancreas using an environmental friendly polyelectrolyte: Alginate
BioChemistry: An Indian Journal (2015)
Rausch C, et al.