Merck
  • The beta-lactoglobulin content of bovine milk: Development and application of a biosensor immunoassay
The beta-lactoglobulin content of bovine milk: Development and application of a biosensor immunoassay
International dairy journal (2017)
Indyk HE, et al.