Merck
  • Sorafenib promotes graft-versus-leukemia activity in mice and humans through IL-15 production in FLT3-ITD-mutant leukemia cells.
Sorafenib promotes graft-versus-leukemia activity in mice and humans through IL-15 production in FLT3-ITD-mutant leukemia cells.
Nature medicine (2018-02-13)
Nimitha R Mathew, Francis Baumgartner, Lukas Braun, David O'Sullivan, Simone Thomas, Miguel Waterhouse, Tony A Müller, Kathrin Hanke, Sanaz Taromi, Petya Apostolova, Anna L Illert, Wolfgang Melchinger, Sandra Duquesne, Annette Schmitt-Graeff, Lena Osswald, Kai-Li Yan, Arnim Weber, Sonia Tugues, Sabine Spath, Dietmar Pfeifer, Marie Follo, Rainer Claus, Michael Lübbert, Christoph Rummelt, Hartmut Bertz, Ralph Wäsch, Johanna Haag, Andrea Schmidts, Michael Schultheiss, Dominik Bettinger, Robert Thimme, Evelyn Ullrich, Yakup Tanriver, Giang Lam Vuong, Renate Arnold, Philipp Hemmati, Dominik Wolf, Markus Ditschkowski, Cordula Jilg, Konrad Wilhelm, Christian Leiber, Sabine Gerull, Jörg Halter, Claudia Lengerke, Thomas Pabst, Thomas Schroeder, Guido Kobbe, Wolf Rösler, Soroush Doostkam, Stephan Meckel, Kathleen Stabla, Stephan K Metzelder, Sebastian Halbach, Tilman Brummer, Zehan Hu, Joern Dengjel, Björn Hackanson, Christoph Schmid, Udo Holtick, Christof Scheid, Alexandros Spyridonidis, Friedrich Stölzel, Rainer Ordemann, Lutz P Müller, Flore Sicre-de-Fontbrune, Gabriele Ihorst, Jürgen Kuball, Jan E Ehlert, Daniel Feger, Eva-Maria Wagner, Jean-Yves Cahn, Jacqueline Schnell, Florian Kuchenbauer, Donald Bunjes, Ronjon Chakraverty, Simon Richardson, Saar Gill, Nicolaus Kröger, Francis Ayuk, Luca Vago, Fabio Ciceri, Antonia M Müller, Takeshi Kondo, Takanori Teshima, Susan Klaeger, Bernhard Kuster, Dennis Dong Hwan Kim, Daniel Weisdorf, Walter van der Velden, Daniela Dörfel, Wolfgang Bethge, Inken Hilgendorf, Andreas Hochhaus, Geoffroy Andrieux, Melanie Börries, Hauke Busch, John Magenau, Pavan Reddy, Myriam Labopin, Joseph H Antin
ZUSAMMENFASSUNG
Individuals with acute myeloid leukemia (AML) harboring an internal tandem duplication (ITD) in the gene encoding Fms-related tyrosine kinase 3 (FLT3) who relapse after allogeneic hematopoietic cell transplantation (allo-HCT) have a 1-year survival rate below 20%. We observed that sorafenib, a multitargeted tyrosine kinase inhibitor, increased IL-15 production by FLT3-ITD+ leukemia cells. This synergized with the allogeneic CD8+ T cell response, leading to long-term survival in six mouse models of FLT3-ITD+ AML. Sorafenib-related IL-15 production caused an increase in CD8+CD107a+IFN-γ+ T cells with features of longevity (high levels of Bcl-2 and reduced PD-1 levels), which eradicated leukemia in secondary recipients. Mechanistically, sorafenib reduced expression of the transcription factor ATF4, thereby blocking negative regulation of interferon regulatory factor 7 (IRF7) activation, which enhanced IL-15 transcription. Both IRF7 knockdown and ATF4 overexpression in leukemia cells antagonized sorafenib-induced IL-15 production in vitro. Human FLT3-ITD+ AML cells obtained from sorafenib responders following sorafenib therapy showed increased levels of IL-15, phosphorylated IRF7, and a transcriptionally active IRF7 chromatin state. The mitochondrial spare respiratory capacity and glycolytic capacity of CD8+ T cells increased upon sorafenib treatment in sorafenib responders but not in nonresponders. Our findings indicate that the synergism of T cells and sorafenib is mediated via reduced ATF4 expression, causing activation of the IRF7-IL-15 axis in leukemia cells and thereby leading to metabolic reprogramming of leukemia-reactive T cells in humans. Therefore, sorafenib treatment has the potential to contribute to an immune-mediated cure of FLT3-ITD-mutant AML relapse, an otherwise fatal complication after allo-HCT.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tandutinib hydrochloride, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.