Merck
  • Protocol for the preparation of Arabidopsis meiotic chromosome spreads and fluorescent in situ hybridization
Protocol for the preparation of Arabidopsis meiotic chromosome spreads and fluorescent in situ hybridization
The Plant Journal (2014)
Bolanos-Villegas P, et al.