Merck
  • Simple efficient methods for the isolation of malate dehydrogenase from thermophilic and mesophilic bacteria.
Simple efficient methods for the isolation of malate dehydrogenase from thermophilic and mesophilic bacteria.
The Biochemical journal (1979-02-01)
I P Wright, T K Sundaram
PMID435244
ZUSAMMENFASSUNG
Malate dehydrogenase from a number of bacteria drawn from several genera and representing the mesophilic, moderately thermophilic and extremely thermophilic classes was isolated by procedures which involve only a small number of steps (in most cases only two), of which the key one is affinity chromatography on 5'-AMP--Sepharose and/or on NAD+--hexane--agarose. Electrophoretic analysis of the native enzymes in polyacrylamide gel and of the denaturated enzymes in sodium dodecyl sulphate/polyacrylamide gel revealed no significant protein impurity in the purified preparations. The yields ranged from about 40% to over 80%. The malate dehydrogenases from the extreme thermophiles and from some of the moderate thermophiles are appreciably less efficient catalytically than their mesophilic homologues.