Merck
  • Effect of propolis and zinc methionine supplementation on improvement of productive, reproductive and immunity performance of local developed in sash strain under egyptian summer conditions.
Effect of propolis and zinc methionine supplementation on improvement of productive, reproductive and immunity performance of local developed in sash strain under egyptian summer conditions.
Egyptian Poultry Science Journal (2019)
Habeib AA, et al.