Merck
  • A randomized, double-blind study of amorolfine 5\% nail lacquer with oral fluconazole compared with oral fluconazole alone in the treatment of fingernail onychomycosis
A randomized, double-blind study of amorolfine 5\% nail lacquer with oral fluconazole compared with oral fluconazole alone in the treatment of fingernail onychomycosis
Indian Journal of Dermatology (2019)
Chandra S, et al.