Merck
  • Cathepsin L1, the Major Protease Involved in Liver Fluke (Fasciola hepatica) virulence propeptide cleavage sites and autoactivation of the zymogen secreted from gastrodermal cells
Cathepsin L1, the Major Protease Involved in Liver Fluke (Fasciola hepatica) virulence propeptide cleavage sites and autoactivation of the zymogen secreted from gastrodermal cells
Test (2004)
Collins PR, et al.