Merck
  • Increased expression and activity of nuclear cathepsin L in cancer cells suggests a novel mechanism of cell transformation
Increased expression and activity of nuclear cathepsin L in cancer cells suggests a novel mechanism of cell transformation
Molecular Cancer Research (2007)
Goulet B, et al.