Merck
  • The multiple functions of cytochrome c and their regulation in life and death decisions of the mammalian cell: from respiration to apoptosis
The multiple functions of cytochrome c and their regulation in life and death decisions of the mammalian cell: from respiration to apoptosis
Mitochondrion (2011)
Huttemann M, et al.