Merck
  • ?-Selective Ribofuranosylation of Alcohols with Ribofuranosyl Iodides and Triphenylphosphine Oxide
?-Selective Ribofuranosylation of Alcohols with Ribofuranosyl Iodides and Triphenylphosphine Oxide
The Journal of Organic Chemistry (2014)
Oka N, et al.