Merck
  • Determination of rotenone residues in raw honey by solid-phase extraction and high-performance liquid chromatography
Determination of rotenone residues in raw honey by solid-phase extraction and high-performance liquid chromatography
Journal of Chromatography A (2000)
Jimenez.JJ, et al.